accessibility menu, dialog, popup

UserName

CAS RN 1801-72-5

H2CNHONHCH2

CAS RN 1801-72-5

IUPAC Name: 1,3-bis(prop-2-en-1-yl)urea
Synonyms: diallyl urea
Molecular Weight (g/mol): 140.19
Molecular Formula: C7H12N2O
InChi Key: QRWVOJLTHSRPOA-UHFFFAOYSA-N
SMILES: C=CCNC(=O)NCC=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results