missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diacetoxybenzene 98.0+%, TCI America™

Catalog No. p-7135673
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R004625G 25 g
R0046500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R004625G Supplier TCI America Supplier No. R004625G
Only null left

Chemical Identifiers

CAS 108-58-7
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00008701
InChI Key STOUHHBZBQBYHH-UHFFFAOYSA-N
Synonym 1,3-diacetoxybenzene, resorcinol diacetate, 1,3-benzenediol, diacetate, m-phenylenediacetate, resorcinol, diacetate, m-phenylene di acetate, 1,3-dihydroxybenzene diacetate, dihydroxybenzene diacetate, 1,3-phenylene diacetate, 1,3-benzenediol, 1,3-diacetate
PubChem CID 7942
IUPAC Name (3-acetyloxyphenyl) acetate
SMILES CC(=O)OC1=CC(=CC=C1)OC(=O)C

Specifications

Color Yellow
Boiling Point 278°C
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diacetoxybenzene
RTECSNumber VH0580000
TSCA Yes