missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-tert-butylbenzene 98.0+%, TCI America™

Catalog No. p-7127295
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
5 mL
Catalog No. Quantity
D24311ML 1 mL
D24315ML 5 mL
2 options

Catalog No. D24311ML

Supplier: TCI America D24311ML

Only null left

Chemical Identifiers

CAS 1014-60-4
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00008830
InChI Key ILNDSSCEZZFNGE-UHFFFAOYSA-N
Synonym 1,3-di-tert-butylbenzene, benzene, 1,3-bis 1,1-dimethylethyl, benzene, m-di-tert-butyl, m-di-tert-butylbenzene, 1,3-di-t-butylbenzene, 1,3-ditert-butyl-benzene, 1,3-ditertiarybutylbenzene, 1,3-di-tert-butyl-benzene, acmc-1bo58, 1,3-bis tert-butyl benzene
PubChem CID 136810
IUPAC Name 1,3-di-tert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC=C1)C(C)(C)C

Specifications

Melting Point 11°C
Color Colorless
Boiling Point 107°C
Quantity 1 mL
Formula Weight 190.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Di-tert-butylbenzene
EINECSNumber (3)-0025
TSCA No