missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-tert-butylbenzene, 99%

CAS: 1014-60-4 | C14H22 | 190.33 g/mol

Supplier:  Thermo Scientific Chemicals B2029006

 View more versions of this product

Catalog No. AAB2029006


Only null left
Explore available promotions

Description

Description

1,3-Di-tert-butylbenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Di-tert-butylbenzene is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

1,3-Di-tert-butylbenzene
10°C to 11°C
106°C to 107°C (18 mmHg)
C14H22
MFCD00008830
1,3-di-tert-butylbenzene, benzene, 1,3-bis 1,1-dimethylethyl, benzene, m-di-tert-butyl, m-di-tert-butylbenzene, 1,3-di-t-butylbenzene, 1,3-ditert-butyl-benzene, 1,3-ditertiarybutylbenzene, 1,3-di-tert-butyl-benzene, acmc-1bo58, 1,3-bis tert-butyl benzene
ILNDSSCEZZFNGE-UHFFFAOYSA-N
1,3-ditert-butylbenzene
136810
99%
1014-60-4
0.859
83°C (183°F)
1.488
5 g
Insoluble in water.
CC(C)(C)C1=CC(=CC=C1)C(C)(C)C
190.33
190.33

Chemical Identifiers

1014-60-4
190.33
ILNDSSCEZZFNGE-UHFFFAOYSA-N
136810
CC(C)(C)C1=CC(=CC=C1)C(C)(C)C
C14H22
MFCD00008830
1,3-di-tert-butylbenzene, benzene, 1,3-bis 1,1-dimethylethyl, benzene, m-di-tert-butyl, m-di-tert-butylbenzene, 1,3-di-t-butylbenzene, 1,3-ditert-butyl-benzene, 1,3-ditertiarybutylbenzene, 1,3-di-tert-butyl-benzene, acmc-1bo58, 1,3-bis tert-butyl benzene
1,3-ditert-butylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only