missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-tert-butylbenzene, 99%

Catalog No. AAB2029003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2029003 1 g
AAB2029006 5 g
AAB2029014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2029003 Supplier Thermo Scientific Chemicals Supplier No. B2029003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1014-60-4 | C14H22 | 190.33 g/mol

1,3-Di-tert-butylbenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Di-tert-butylbenzene is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 1014-60-4
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00008830
InChI Key ILNDSSCEZZFNGE-UHFFFAOYSA-N
Synonym 1,3-di-tert-butylbenzene, benzene, 1,3-bis 1,1-dimethylethyl, benzene, m-di-tert-butyl, m-di-tert-butylbenzene, 1,3-di-t-butylbenzene, 1,3-ditert-butyl-benzene, 1,3-ditertiarybutylbenzene, 1,3-di-tert-butyl-benzene, acmc-1bo58, 1,3-bis tert-butyl benzene
PubChem CID 136810
IUPAC Name 1,3-ditert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC=C1)C(C)(C)C

Specifications

Melting Point 10°C to 11°C
Density 0.859
Boiling Point 106°C to 107°C (18 mmHg)
Flash Point 83°C (183°F)
Refractive Index 1.488
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 190.33
Percent Purity 99%
Chemical Name or Material 1,3-Di-tert-butylbenzene
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only