missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di(p-tolyl)thiourea 98.0+%, TCI America™

Catalog No. D08035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08035G 5 g
D080325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D08035G Supplier TCI America Supplier No. D08035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 621-01-2
Molecular Formula C15H16N2S
Molecular Weight (g/mol) 256.367
MDL Number MFCD00008541
InChI Key ULNVBRUIKLYGDF-UHFFFAOYSA-N
PubChem CID 688021
IUPAC Name 1,3-bis(4-methylphenyl)thiourea
SMILES CC1=CC=C(C=C1)NC(=S)NC2=CC=C(C=C2)C

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 5 g
Formula Weight 256.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di(p-tolyl)thiourea
RTECSNumber FE0875000
TSCA No