missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di(o-tolyl)thiourea 98.0+%, TCI America™

Catalog No. D080225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D080225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D080225G Supplier TCI America Supplier No. D080225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137-97-3
Molecular Formula C15H16N2S
Molecular Weight (g/mol) 256.37
MDL Number MFCD00025922
InChI Key KWPNNZKRAQDVPZ-UHFFFAOYSA-N
Synonym di-o-tolylthiourea, 1,3-di o-tolyl thiourea, 1,3-di-o-tolylthiourea, n,n'-di-o-tolylthiourea, 1,3-di-o-tolyl-2-thiourea, di-o-toluylthiourea, 1,3-bis o-tolyl thiourea, n,n'-bis 2-methylphenyl thiourea, 2,2'-dimethylthiocarbanilide, usaf ek-1651
PubChem CID 688028
IUPAC Name 1,3-bis(2-methylphenyl)thiourea
SMILES CC1=CC=CC=C1NC(=S)NC1=CC=CC=C1C

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 25 g
Formula Weight 256.37
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di(o-tolyl)thiourea
EINECSNumber (3)-2206
RTECSNumber FE0700000
TSCA Yes