missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di(2-pyridyl)propane 97.0+%, TCI America™

Catalog No. D5415500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5415500G 500 g
D541525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5415500G Supplier TCI America Supplier No. D5415500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15937-81-2
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
InChI Key UHPWYBATPHCMGD-UHFFFAOYSA-N
PubChem CID 322131
IUPAC Name 2-(3-pyridin-2-ylpropyl)pyridine
SMILES C1=CC=NC(=C1)CCCC2=CC=CC=N2

Specifications

Boiling Point 117°C
Quantity 500 g
Formula Weight 198.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Di(2-pyridyl)propane