missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di(2-pyridyl)-1,3-propanedione 98.0+%, TCI America™

Catalog No. p-7127342
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25121G 1 g
D25125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25121G Supplier TCI America Supplier No. D25121G
Only null left

Chemical Identifiers

CAS 10198-89-7
Molecular Formula C13H10N2O2
Molecular Weight (g/mol) 226.235
MDL Number MFCD01321157
InChI Key DCGUVLMWGIPVDP-UHFFFAOYSA-N
Synonym Dipicolinoylmethane
PubChem CID 2728494
IUPAC Name 1,3-dipyridin-2-ylpropane-1,3-dione
SMILES C1=CC=NC(=C1)C(=O)CC(=O)C2=CC=CC=N2

Specifications

Melting Point 106°C
Color Yellow
Quantity 1 g
Formula Weight 226.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di(2-pyridyl)-1,3-propanedione