accessibility menu, dialog, popup

UserName

NOON

CAS RN 10198-89-7

IUPAC Name: 1,3-bis(pyridin-2-yl)propane-1,3-dione
Synonyms: 1,3-bis(pyridin-2-yl)propane-1,3-dione
Molecular Weight (g/mol): 226.24
Molecular Formula: C13H10N2O2
InChi Key: DCGUVLMWGIPVDP-UHFFFAOYSA-N
SMILES: O=C(CC(=O)C1=CC=CC=N1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results