missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di(1-adamantyl)imidazolinium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. d3620500mg
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3620500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3620500MG Supplier TCI America Supplier No. D3620500MG
Only null left

Chemical Identifiers

CAS 1176202-63-3
Molecular Formula C23H37BF4N2
Molecular Weight (g/mol) 428.367
InChI Key AFLPPSBVXPAHAH-UHFFFAOYSA-O
PubChem CID 56923593
IUPAC Name 1,3-bis(1-adamantyl)imidazolidin-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1CN(C[NH+]1C23CC4CC(C2)CC(C4)C3)C56CC7CC(C5)CC(C7)C6

Specifications

Quantity 500 mg
UN Number 1759
Formula Weight 426.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di(1-adamantyl)imidazolinium Tetrafluoroborate