missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclooctadiene (stabilized with TBC) 95.0+%, TCI America™

Catalog No. c0502500ml
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0502500ML 500 mL
C050225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0502500ML Supplier TCI America Supplier No. C0502500ML

Chemical Identifiers

CAS 1700-10-3
Molecular Formula C8H12
Molecular Weight (g/mol) 108.18
MDL Number MFCD00001751
InChI Key RRKODOZNUZCUBN-CCAGOZQPSA-N
Synonym 1,3-cyclooctadiene, cyclooctadiene, 1z,3z-cycloocta-1,3-diene, cis,cis-1,3-cyclooctadiene, cycloocta-1,3-diene, 1,3-cyclooctadiene, z,z, cyclooctadienes, 1.3-cyclooctadiene, 1e,3z-1,3-cyclooctadiene, 1,3-cod
PubChem CID 299882
IUPAC Name (1Z,3Z)-cycloocta-1,3-diene
SMILES C1CC\C=C/C=C\C1

Specifications

Color Yellow
Boiling Point 143°C
Quantity 500 mL
UN Number 2520
Formula Weight 108.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Cyclooctadiene (stabilized with TBC)
EINECSNumber (3)-2232
TSCA Yes