accessibility menu, dialog, popup

UserName

CAS RN 1700-10-3

IUPAC Name: (1Z,3Z)-cycloocta-1,3-diene
Synonyms: (1Z,3Z)-cycloocta-1,3-diene
Molecular Weight (g/mol): 108.18
Molecular Formula: C8H12
InChi Key: RRKODOZNUZCUBN-CCAGOZQPSA-N
SMILES: C1CC\C=C/C=C\C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)