missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Cyclooctadiene, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -53°C |
| Density | 0.869 |
| Boiling Point | 142°C to 144°C |
| Flash Point | 24°C (75°F) |
| Odor | Characteristic |
| Refractive Index | 1.493 |
| Quantity | 5 g |
| UN Number | UN2520 |
| Beilstein | 1901033 |
| Formula Weight | 108.18 |
| Show More |
Safety and Handling
| Hazard Category | H226-H304-H315-H319-H335 |
| Hazard Statement | GHS H Statement H304-H226-H315-H319-H335 May be fatal if swallowed and enters airways. Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOOCTADIENES |
| EINECSNumber | 216-929-1 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only