missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanediamine (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. c08135ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
C08135ML 5 mL
C081325ML 25 mL
2 options

Catalog No. C08135ML

Supplier: TCI America C08135ML

Only null left

Chemical Identifiers

CAS 3385-21-5
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00059563
InChI Key GEQHKFFSPGPGLN-UHFFFAOYSA-N
Synonym 1,3-diaminocyclohexane, 1,3-cyclohexanediamine, cyclohex-1,3-ylenediamine, ccris 6679, 2,4-diaminocyclohexane, acmc-20ap4j, 1,3-diamino-cyclohexane, cyclohexane-1.3-diamine, rac-1,3-cyclohexanediamine, 1r,3s-3-azaniumylcyclohexyl azanium
PubChem CID 18814
IUPAC Name cyclohexane-1,3-diamine
SMILES C1CC(CC(C1)N)N

Specifications

Color Yellow
Boiling Point 194°C
Quantity 5 mL
UN Number 2735
Formula Weight 114.19
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,3-Cyclohexanediamine (cis- and trans- mixture)
EINECSNumber (3)-3871
RTECSNumber GU8750000
TSCA Yes