missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Cyclohexanediamine (cis- and trans- mixture) 95.0+%, TCI America™
Supplier: TCI America C081325ML
Specifications
| 1,3-Cyclohexanediamine (cis- and trans- mixture) | |
| Yellow | |
| C6H14N2 | |
| 25 mL | |
| 1,3-diaminocyclohexane, 1,3-cyclohexanediamine, cyclohex-1,3-ylenediamine, ccris 6679, 2,4-diaminocyclohexane, acmc-20ap4j, 1,3-diamino-cyclohexane, cyclohexane-1.3-diamine, rac-1,3-cyclohexanediamine, 1r,3s-3-azaniumylcyclohexyl azanium | |
| C1CC(CC(C1)N)N | |
| 114.192 | |
| 114.19 | |
| Liquid |
| 3385-21-5 | |
| 194°C | |
| MFCD00059563 | |
| 2735 | |
| GEQHKFFSPGPGLN-UHFFFAOYSA-N | |
| cyclohexane-1,3-diamine | |
| 18814 | |
| ≥95.0% (GC,T) |
Chemical Identifiers
| 3385-21-5 | |
| 114.192 | |
| GEQHKFFSPGPGLN-UHFFFAOYSA-N | |
| 18814 | |
| C1CC(CC(C1)N)N |
| C6H14N2 | |
| MFCD00059563 | |
| 1,3-diaminocyclohexane, 1,3-cyclohexanediamine, cyclohex-1,3-ylenediamine, ccris 6679, 2,4-diaminocyclohexane, acmc-20ap4j, 1,3-diamino-cyclohexane, cyclohexane-1.3-diamine, rac-1,3-cyclohexanediamine, 1r,3s-3-azaniumylcyclohexyl azanium | |
| cyclohexane-1,3-diamine |
Safety and Handling
EINECSNumber : (3)-3871
RTECSNumber : GU8750000
TSCA : Yes