missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chloro-2-pyridyl)piperazine, 98%

CAS: 87394-55-6 | C9H12ClN3 | 197.666 g/mol

Supplier:  Thermo Scientific Chemicals H5085903

 View more versions of this product

Catalog No. AAH5085903


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-(3-Chloro-2-pyridyl)piperazine
C9H12ClN3
1 g
HLCPXCHNWZGOMT-UHFFFAOYSA-N
1-(3-chloropyridin-2-yl)piperazine
11412944
98%
87394-55-6
MFCD08061034
1-3-chloropyridin-2-yl piperazine, 1-3-chloro-2-pyridyl piperazine, 1-3-chloro-pyridin-2-yl-piperazine, 1-3-chloro-2-pyridinyl piperazine, acmc-20apbr, 3-chloro-2-pyridyl piperazine, n 3-chloropyridin-2-yl-piperazine, n-3-chloropyridin-2-yl-piperazine, piperazine,1-3-chloro-2-pyridinyl
C1CN(CCN1)C2=C(C=CC=N2)Cl
197.666
197.67

Chemical Identifiers

87394-55-6
197.666
HLCPXCHNWZGOMT-UHFFFAOYSA-N
11412944
C1CN(CCN1)C2=C(C=CC=N2)Cl
C9H12ClN3
MFCD08061034
1-3-chloropyridin-2-yl piperazine, 1-3-chloro-2-pyridyl piperazine, 1-3-chloro-pyridin-2-yl-piperazine, 1-3-chloro-2-pyridinyl piperazine, acmc-20apbr, 3-chloro-2-pyridyl piperazine, n 3-chloropyridin-2-yl-piperazine, n-3-chloropyridin-2-yl-piperazine, piperazine,1-3-chloro-2-pyridinyl
1-(3-chloropyridin-2-yl)piperazine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions

RUO – Research Use Only