missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chloro-2-pyridyl)piperazine, 98%

Catalog No. p-7033706
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50859MD 250 mg
AAH5085903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50859MD Supplier Thermo Scientific Chemicals Supplier No. H50859MD
Only null left

CAS: 87394-55-6 | C9H12ClN3 | 197.666 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87394-55-6
Molecular Formula C9H12ClN3
Molecular Weight (g/mol) 197.666
MDL Number MFCD08061034
InChI Key HLCPXCHNWZGOMT-UHFFFAOYSA-N
Synonym 1-3-chloropyridin-2-yl piperazine, 1-3-chloro-2-pyridyl piperazine, 1-3-chloro-pyridin-2-yl-piperazine, 1-3-chloro-2-pyridinyl piperazine, acmc-20apbr, 3-chloro-2-pyridyl piperazine, n 3-chloropyridin-2-yl-piperazine, n-3-chloropyridin-2-yl-piperazine, piperazine,1-3-chloro-2-pyridinyl
PubChem CID 11412944
IUPAC Name 1-(3-chloropyridin-2-yl)piperazine
SMILES C1CN(CCN1)C2=C(C=CC=N2)Cl

Specifications

Quantity 250 mg
Formula Weight 197.67
Percent Purity 98%
Chemical Name or Material 1-(3-Chloro-2-pyridyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only