missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chloro-2-hydroxyphenyl)ethan-1-one, 97%, Thermo Scientific™

Catalog No. p-3484591
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB10071CB 250mg Amber Glass Bottle
BTB10071DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB10071CB Supplier Thermo Scientific Chemicals Supplier No. BTB10071CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3226-34-4
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00100492
InChI Key GBWVDQBTXFIIJF-UHFFFAOYSA-N
Synonym 1-3-chloro-2-hydroxyphenyl ethanone, 1-3-chloro-2-hydroxyphenyl ethan-1-one, 3'-chloro-2'-hydroxyacetophenone, ethanone,1-3-chloro-2-hydroxyphenyl, 2'-hydroxy-3'-chloroacetophenone, 2'-hydroxy-3'-chloro acetophenone, 1-3-chloro-2-hydroxyphenyl-ethanone, 1-3-chloro-2-hydroxy-phenyl-ethanone, 1-3-chloro-2-hydroxyphenyl ethanone #, 2 inverted exclamation mark-hydroxy-3-chloroacetophenone
PubChem CID 593414
IUPAC Name 1-(3-chloro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C(Cl)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 170.59
Percent Purity 97%
Grade Technical
Chemical Name or Material 1-(3-Chloro-2-hydroxyphenyl)ethan-1-one
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.