CAS RN 3226-34-4
IUPAC Name:
1-(3-chloro-2-hydroxyphenyl)ethan-1-one
Synonyms:
1-(3-chloro-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol):
170.59
Molecular Formula:
C8H7ClO2
InChi Key:
GBWVDQBTXFIIJF-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(O)C(Cl)=CC=C1
1
–
1
of
1
results
1
1-(3-Chloro-2-hydroxyphenyl)ethan-1-one, 97%, Thermo Scientific™
CAS: 3226-34-4 Molecular Formula: C8H7ClO2 Molecular Weight (g/mol): 170.59 MDL Number: MFCD00100492 InChI Key: GBWVDQBTXFIIJF-UHFFFAOYSA-N Synonym: 1-3-chloro-2-hydroxyphenyl ethanone,1-3-chloro-2-hydroxyphenyl ethan-1-one,3'-chloro-2'-hydroxyacetophenone,ethanone,1-3-chloro-2-hydroxyphenyl,2'-hydroxy-3'-chloroacetophenone,2'-hydroxy-3'-chloro acetophenone,1-3-chloro-2-hydroxyphenyl-ethanone,1-3-chloro-2-hydroxy-phenyl-ethanone,1-3-chloro-2-hydroxyphenyl ethanone #,2 inverted exclamation mark-hydroxy-3-chloroacetophenone PubChem CID: 593414 IUPAC Name: 1-(3-chloro-2-hydroxyphenyl)ethanone SMILES: CC(=O)C1=C(O)C(Cl)=CC=C1