missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Butanediol 99.0+%, TCI America™
Supplier: TCI America B377025G
Specifications
| 1,3-Butanediol [for Biochemical Research] | |
| Yellow | |
| C4H10O2 | |
| 25 g | |
| PUPZLCDOIYMWBV-UHFFFAOYSA-N | |
| butane-1,3-diol | |
| 7896 | |
| 90.12 | |
| Liquid |
| 107-88-0 | |
| 207°C | |
| MFCD00004554 | |
| 1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol | |
| CC(CCO)O | |
| 90.122 | |
| CHEBI:52683 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 107-88-0 | |
| 90.122 | |
| PUPZLCDOIYMWBV-UHFFFAOYSA-N | |
| 7896 | |
| butane-1,3-diol |
| C4H10O2 | |
| MFCD00004554 | |
| 1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol | |
| CHEBI:52683 | |
| CC(CCO)O |
Safety and Handling
EINECSNumber : (2)-0235
RTECSNumber : EK0440000
TSCA : Yes