missing translation for 'onlineSavingsMsg'
Learn More

1,3-Butanediol Dimethacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. p-7118829
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B106525ML 25 mL
B1065500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B106525ML Supplier TCI America Supplier No. B106525ML

Chemical Identifiers

CAS 1189-08-8
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.272
MDL Number MFCD00014930
InChI Key VDYWHVQKENANGY-UHFFFAOYSA-N
Synonym 1,3-Butylene Glycol Dimethacrylate
PubChem CID 70916
IUPAC Name 3-(2-methylprop-2-enoyloxy)butyl 2-methylprop-2-enoate
SMILES CC(CCOC(=O)C(=C)C)OC(=O)C(=C)C

Specifications

Color Colorless
Boiling Point 290°C
Quantity 25 mL
Formula Weight 226.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Butanediol Dimethacrylate (stabilized with MEHQ)
EINECSNumber (2)-0958&(2)-1058&(2)-1059
TSCA Yes