CAS RN 1189-08-8
IUPAC Name:
4-[(2-methylprop-2-enoyl)oxy]butan-2-yl 2-methylprop-2-enoate
Synonyms:
4-[(2-methylprop-2-enoyl)oxy]butan-2-yl 2-methylprop-2-enoate
Molecular Weight (g/mol):
226.27
Molecular Formula:
C12H18O4
InChi Key:
VDYWHVQKENANGY-UHFFFAOYNA-N
SMILES:
CC(CCOC(=O)C(C)=C)OC(=O)C(C)=C
1
–
1
of
1
results
1
1,3-Butanediol Dimethacrylate (stabilized with MEHQ) 98.0+%, TCI America™
CAS: 1189-08-8 Molecular Formula: C12H18O4 Molecular Weight (g/mol): 226.272 MDL Number: MFCD00014930 InChI Key: VDYWHVQKENANGY-UHFFFAOYSA-N Synonym: 1,3-Butylene Glycol Dimethacrylate PubChem CID: 70916 IUPAC Name: 3-(2-methylprop-2-enoyloxy)butyl 2-methylprop-2-enoate SMILES: CC(CCOC(=O)C(=C)C)OC(=O)C(=C)C