missing translation for 'onlineSavingsMsg'
Learn More

1,3-Butanediol, 99%, Spectrum™ Chemical

Catalog No. 18608056
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608056 500 mL Glass Bottle
18608052 2.5 L Glass Bottle
18608055 4 L Glass Bottle
18608053 20 L Glass Bottle
18608054 200 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608056 Supplier Spectrum Chemical Mfg Cor Supplier No. B2147500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 107-88-0

Specifications

CAS Max % 1
Boiling Point 204°C to 207.5°C (399.2°F to 405.5°F)
Packaging Glass Bottle
Quantity 500 mL
Molecular Weight (g/mol) 90.12
Percent Purity 0.99
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material 1,3-Butanediol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.