missing translation for 'onlineSavingsMsg'
Learn More

(±)-1,3-Butanediol, 99%, extra pure

CAS: 107-88-0 | C4H10O2 | 90.12 g/mol

Supplier:  Thermo Scientific Chemicals 107620100

 View more versions of this product

Catalog No. AC107620100


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(±)-1,3-Butanediol
98.5
-54.0°C
1.0100g/mL
109°C
Authentic
Plastic drum
1.4385 to 1.4405
MFCD00004554
01, 477
15, 1570
Solubility in water: soluble. Other solubilities: soluble in acetone, methyl ethyl ketone, ethanol, slightly soluble in ether, practically insoluble in aliphatic hydrocarbons,, benzene, toluene and ccl4.
CC(CCO)O
90.12
24.6 mm2/s (50°C)
90.12
Extra Pure
107-88-0
100.0
Brown to Colorless
203.0°C to 204.0°C
0.05% max.
98.5% min. (GC)
C4H10O2
CH3CH(OH)CH2CH2OH
10 L
1.01
1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol
PUPZLCDOIYMWBV-UHFFFAOYSA-N
butane-1,3-diol
7896
CHEBI:52683
99%
Liquid

Chemical Identifiers

107-88-0
90.12
PUPZLCDOIYMWBV-UHFFFAOYSA-N
7896
butane-1,3-diol
C4H10O2
MFCD00004554
1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol
CHEBI:52683
CC(CCO)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-529-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only