missing translation for 'onlineSavingsMsg'
Learn More

(±)-1,3-Butanediol, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012418
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012418 1 mL
111012419 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012418 Supplier Merck Emd Millipore Supplier No. 090681ML
Only null left

(+/-)-1,3-Butanediol is an organic alcohol with antimicrobial activity against a wide variety of pathogens.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Ignition Point 741°F
CAS 107-88-0
Vapor Pressure 0.06 mmHg (20°C)
Vapor Density 3.1 (20°C, vs. air)
Density 1.005 g/mL (at 25°C)
Flash Point 108°C
Linear Formula CH3 CH(OH)CH2 CH2 OH
MDL Number MFCD00004554
Boiling Point 203°C to 204°C
Synonym 1,3-Butylene glycol
Type Analytical Standard
Quantity 1 mL
Formula Weight 90.12
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less