missing translation for 'onlineSavingsMsg'
Learn More

1,3-Butadiene Monoepoxide 97.0+%, TCI America™

Catalog No. p-7128945
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03371G 1 g
E03375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03371G Supplier TCI America Supplier No. E03371G
Only null left

Chemical Identifiers

CAS 930-22-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00005149
InChI Key GXBYFVGCMPJVJX-UHFFFAOYSA-N
Synonym butadiene monoxide, oxirane, ethenyl, 3,4-epoxy-1-butene, 2-vinyloxirane, butadiene epoxide, 1,2-epoxy-3-butene, vinyloxirane, butadiene oxide, butadiene monoepoxide, butadiene monooxide
PubChem CID 13586
IUPAC Name 2-ethenyloxirane
SMILES C=CC1CO1

Specifications

Melting Point -135°C
Color Yellow
Boiling Point 67°C
Quantity 1 g
UN Number 1993
Formula Weight 70.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Butadiene Monoepoxide
RTECSNumber EM7350000
TSCA Yes