missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(vinylsulfonyl)-2-propanol 98.0+%, TCI America™

Catalog No. p-7119235
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17451G 1 g
B17455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17451G Supplier TCI America Supplier No. B17451G

Chemical Identifiers

CAS 67006-32-0
Molecular Formula C7H12O5S2
Molecular Weight (g/mol) 240.288
MDL Number MFCD00671498
InChI Key SOBDFTUDYRPGJY-UHFFFAOYSA-N
PubChem CID 13684924
IUPAC Name 1,3-bis(ethenylsulfonyl)propan-2-ol
SMILES C=CS(=O)(=O)CC(CS(=O)(=O)C=C)O

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 1 g
Formula Weight 240.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(vinylsulfonyl)-2-propanol
EINECSNumber (2)-3134
TSCA No