accessibility menu, dialog, popup

UserName

CAS RN 67006-32-0

IUPAC Name: [3-(ethenesulfonyl)-2-hydroxypropanesulfonyl]ethene
Synonyms: [3-(ethenesulfonyl)-2-hydroxypropanesulfonyl]ethene
Molecular Weight (g/mol): 240.29
Molecular Formula: C7H12O5S2
InChi Key: SOBDFTUDYRPGJY-UHFFFAOYSA-N
SMILES: OC(CS(=O)(=O)C=C)CS(=O)(=O)C=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)