missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[(trimethylsilyl)ethynyl]benzene 98.0+%, TCI America™

Catalog No. B32161G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B32161G Supplier TCI America Supplier No. B32161G
Only null left

Chemical Identifiers

CAS 38170-80-8
Molecular Formula C16H22Si2
Molecular Weight (g/mol) 270.522
MDL Number MFCD00078305
InChI Key ASZOZZMGMNVKDV-UHFFFAOYSA-N
Synonym 1,3-bis trimethylsilyl ethynyl benzene, 1,3-bis trimethylsilylethynyl benzene, trimethyl 2-3-2-trimethylsilyl ethynyl phenyl ethynyl silane, acmc-209iy7, 1,3-bis trimethylsilyl ethynylbenzene, 1,3-bis 2-trimethylsilyl ethynyl benzene, 1,3-bis-trimethylsilyl-ethynyl-benzene, benzene,1,3-bis 2-trimethylsilyl ethynyl, trimethyl-2-3-2-trimethylsilylethynyl phenyl ethynyl silane
PubChem CID 142239
IUPAC Name trimethyl-[2-[3-(2-trimethylsilylethynyl)phenyl]ethynyl]silane
SMILES C[Si](C)(C)C#CC1=CC(=CC=C1)C#C[Si](C)(C)C

Specifications

Melting Point 60°C
Color White-Yellow
Quantity 1 g
Formula Weight 270.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[(trimethylsilyl)ethynyl]benzene
TSCA No
Recommended Storage Refrigerator