missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(trifluoromethyl)benzene 98.0+%, TCI America™

Catalog No. p-7130619
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H058525G 25 g
H0585100G 100 g
H0585500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H058525G Supplier TCI America Supplier No. H058525G
Only null left

Chemical Identifiers

CAS 402-31-3
Molecular Formula C8H4F6
Molecular Weight (g/mol) 214.11
MDL Number MFCD00000392
InChI Key SJBBXFLOLUTGCW-UHFFFAOYSA-N
Synonym 1,3-bis trifluoromethyl benzene, benzene, 1,3-bis trifluoromethyl, 1,3-di trifluoromethyl benzene, 1,3-bis trifluoromethyl-benzene, m-bis trifluoromethyl benzene, m-ditrifluoromethylbenzene, hexafluoro-m-xylene, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-m-xylene, mtf-tfm, a,a,a,a',a',a'-hexafluoro-m-xylene
PubChem CID 223106
IUPAC Name 1,3-bis(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 116°C
Quantity 25 g
UN Number 1993
Formula Weight 214.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Bis(trifluoromethyl)benzene
EINECSNumber (3)-0054
TSCA Yes