missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(tert-butoxycarbonyl)thiourea 98.0+%, TCI America™

Catalog No. B45595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45595G 5 g
B45591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45595G Supplier TCI America Supplier No. B45595G
Only null left

Chemical Identifiers

CAS 145013-05-4
Molecular Formula C11H20N2O4S
Molecular Weight (g/mol) 276.351
MDL Number MFCD03093917
InChI Key CSOJECDGWHHWRS-UHFFFAOYSA-N
Synonym 1,3-Di-Boc-thiourea
PubChem CID 5193990
IUPAC Name tert-butyl N-[(2-methylpropan-2-yl)oxycarbonylcarbamothioyl]carbamate
SMILES CC(C)(C)OC(=O)NC(=S)NC(=O)OC(C)(C)C

Specifications

Melting Point 132°C
Color White
Quantity 5 g
Formula Weight 276.35
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(tert-butoxycarbonyl)thiourea