accessibility menu, dialog, popup

UserName

CAS RN 145013-05-4

H3CCH3H3COONHSNHOOCH3CH3H3C

CAS RN 145013-05-4

IUPAC Name: tert-butyl N-({[(tert-butoxy)carbonyl]amino}methanethioyl)carbamate
Synonyms: tert-butyl N-{[(tert-butoxycarbonyl)amino]methanethioyl}carbamate
Molecular Weight (g/mol): 276.35
Molecular Formula: C11H20N2O4S
InChi Key: CSOJECDGWHHWRS-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC(=S)NC(=O)OC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results