missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(methylthio)-2-methoxypropane 97.0+%, TCI America™

Catalog No. B17095G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17095G Supplier TCI America Supplier No. B17095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31805-84-2
Molecular Formula C6H14OS2
Molecular Weight (g/mol) 166.297
MDL Number MFCD00026040
InChI Key MOISORXUYSHXRU-UHFFFAOYSA-N
Synonym 1,3-bis methylthio-2-methoxypropane, 2-methoxy-1,3-bis methylsulfanyl propane, 2-methoxy-1,3-bis methylthio propane, 1,3-bis-methylthio-2-methoxypropane, propane, 2-methoxy-1,3-bis methylthio, acmc-1cktt, moisorxuyshxru-uhfffaoysa, propane,2-methoxy-1,3-bis methylthio, 2-methoxy-1,3-bis methylsulfanyl-propane, 2-methoxy-1,3-bis methylsulfanyl propane #
PubChem CID 520544
IUPAC Name 2-methoxy-1,3-bis(methylsulfanyl)propane
SMILES COC(CSC)CSC

Specifications

Color Yellow
Boiling Point 97°C
Quantity 5 g
Formula Weight 166.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Bis(methylthio)-2-methoxypropane