missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(isocyanatomethyl)cyclohexane (cis- and trans- mixture) 99.0+%, TCI America™

Catalog No. p-7119104
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B153825ML 25 mL
B1538500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B153825ML Supplier TCI America Supplier No. B153825ML
Only null left

Chemical Identifiers

CAS 38661-72-2
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.23
MDL Number MFCD00129947
InChI Key XSCLFFBWRKTMTE-UHFFFAOYNA-N
PubChem CID 53172
IUPAC Name 1,3-bis(isocyanatomethyl)cyclohexane
SMILES O=C=NCC1CCCC(CN=C=O)C1

Specifications

Color Colorless
Quantity 25 mL
UN Number 2206
Formula Weight 194.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Bis(isocyanatomethyl)cyclohexane (cis- and trans- mixture)
EINECSNumber (3)-2482
TSCA Yes
Recommended Storage Refrigerator