missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(diphenylphosphino)propane 98.0+%, TCI America™

Catalog No. p-7118874
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B11385G 5 g
B113825G 25 g
2 options

Catalog No. B11385G

Supplier: TCI America B11385G

Only null left

Chemical Identifiers

CAS 6737-42-4
Molecular Formula C27H26P2
Molecular Weight (g/mol) 412.45
MDL Number MFCD00003050
InChI Key LVEYOSJUKRVCCF-UHFFFAOYSA-N
Synonym 1,3-bis diphenylphosphino propane, dppp, trimethylenebis diphenylphosphine, phosphine, 1,3-propanediylbis diphenyl, 3-diphenylphosphanyl propyl diphenylphosphane, 3-diphenylphosphanylpropyl diphenyl phosphane, 1,3-bis diphenyphosphino propane, 1,3-bis diphenylphosphino propane dppp, bis 1,3-diphenylphosphino propane, 1,3-bis-diphenylphosphino propane
PubChem CID 81219
IUPAC Name [3-(diphenylphosphanyl)propyl]diphenylphosphane
SMILES C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 63°C
Color White
Quantity 5 g
Formula Weight 412.45
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(diphenylphosphino)propane