accessibility menu, dialog, popup

UserName

CAS RN 6737-42-4

IUPAC Name: [3-(diphenylphosphanyl)propyl]diphenylphosphane
Synonyms: [3-(diphenylphosphanyl)propyl]diphenylphosphane
Molecular Weight (g/mol): 412.45
Molecular Formula: C27H26P2
InChi Key: LVEYOSJUKRVCCF-UHFFFAOYSA-N
SMILES: C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)