missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[1,3-Bis(diphenylphosphino)propane]palladium(II) Dichloride 98.0+%, TCI America™
$180.81 - $561.08
Chemical Identifiers
| CAS | 59831-02-6 |
|---|---|
| Molecular Formula | C27H26Cl2P2Pd |
| Molecular Weight (g/mol) | 589.77 |
| MDL Number | MFCD03844773 |
| InChI Key | LDFBXJODFADZBN-UHFFFAOYSA-L |
| Synonym | Dichloro[1,3-bis(diphenylphosphino)propane]palladium(II) |
| PubChem CID | 131664225 |
| IUPAC Name | palladium(2+) [3-(diphenylphosphanyl)propyl]diphenylphosphane dichloride |
| SMILES | [Cl-].[Cl-].[Pd++].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 59831-02-6 | |
| 589.77 | |
| LDFBXJODFADZBN-UHFFFAOYSA-L | |
| 131664225 | |
| [Cl-].[Cl-].[Pd++].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1 |
| C27H26Cl2P2Pd | |
| MFCD03844773 | |
| Dichloro[1,3-bis(diphenylphosphino)propane]palladium(II) | |
| palladium(2+) [3-(diphenylphosphanyl)propyl]diphenylphosphane dichloride |
Specifications
| 59831-02-6 | |
| MFCD03844773 | |
| Dichloro[1,3-bis(diphenylphosphino)propane]palladium(II) | |
| [Cl-].[Cl-].[Pd++].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 589.77 | |
| 589.77 | |
| Crystalline Powder |
| C27H26Cl2P2Pd | |
| 1 g | |
| LDFBXJODFADZBN-UHFFFAOYSA-L | |
| palladium(2+) [3-(diphenylphosphanyl)propyl]diphenylphosphane dichloride | |
| 131664225 | |
| ≥98.0% (T) | |
| [1,3-Bis(diphenylphosphino)propane]palladium(II) Dichloride |
Safety and Handling
TSCA : No