accessibility menu, dialog, popup

UserName

CAS RN 59831-02-6

PPClClPd

CAS RN 59831-02-6

IUPAC Name: palladium(2+) [3-(diphenylphosphanyl)propyl]diphenylphosphane dichloride
Synonyms: palladium(2+) [3-(diphenylphosphanyl)propyl]diphenylphosphane dichloride
Molecular Weight (g/mol): 589.77
Molecular Formula: C27H26Cl2P2Pd
InChi Key: LDFBXJODFADZBN-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pd++].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results