missing translation for 'onlineSavingsMsg'
Learn More

[1,3-Bis(diphenylphosphino)propane]palladium(II) Dichloride 98.0+%, TCI America™

Catalog No. B21925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21925G 5 g
B21921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21925G Supplier TCI America Supplier No. B21925G
Only null left

Chemical Identifiers

CAS 59831-02-6
Molecular Formula C27H26Cl2P2Pd
Molecular Weight (g/mol) 589.77
MDL Number MFCD03844773
InChI Key LDFBXJODFADZBN-UHFFFAOYSA-L
Synonym Dichloro[1,3-bis(diphenylphosphino)propane]palladium(II)
PubChem CID 131664225
IUPAC Name palladium(2+) [3-(diphenylphosphanyl)propyl]diphenylphosphane dichloride
SMILES [Cl-].[Cl-].[Pd++].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 589.77
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,3-Bis(diphenylphosphino)propane]palladium(II) Dichloride