missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[1,3-Bis(diphenylphosphino)propane]nickel(II) Dichloride 98.0+%, TCI America™
$59.65 - $203.17
Chemical Identifiers
| CAS | 15629-92-2 |
|---|---|
| Molecular Formula | C27H26Cl2NiP2 |
| Molecular Weight (g/mol) | 542.04 |
| MDL Number | MFCD00015318 |
| InChI Key | REQVOKPBMSCTNI-UHFFFAOYSA-L |
| Synonym | [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II) |
| PubChem CID | 131675641 |
| IUPAC Name | [3-(diphenylphosphanyl)propyl]diphenylphosphane; dichloronickelbis(ylium) |
| SMILES | Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 15629-92-2 | |
| 542.04 | |
| REQVOKPBMSCTNI-UHFFFAOYSA-L | |
| 131675641 | |
| Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1 |
| C27H26Cl2NiP2 | |
| MFCD00015318 | |
| [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II) | |
| [3-(diphenylphosphanyl)propyl]diphenylphosphane; dichloronickelbis(ylium) |
Specifications
| 15629-92-2 | |
| C27H26Cl2NiP2 | |
| 5 g | |
| REQVOKPBMSCTNI-UHFFFAOYSA-L | |
| [3-(diphenylphosphanyl)propyl]diphenylphosphane; dichloronickelbis(ylium) | |
| 131675641 | |
| ≥98.0% (T) | |
| [1,3-Bis(diphenylphosphino)propane]nickel(II) Dichloride |
| 213°C | |
| MFCD00015318 | |
| [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II) | |
| Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 542.04 | |
| 542.05 | |
| Crystalline Powder |
Safety and Handling
TSCA : No