Learn More
[1,3-Bis(diphenylphosphino)propane]nickel(II) chloride, 98+%
CAS: 15629-92-2 | C27H26Cl2NiP2 | 542.04 g/mol
$73.34 - $142.39
Chemical Identifiers
| CAS | 15629-92-2 |
|---|---|
| Molecular Formula | C27H26Cl2NiP2 |
| Molecular Weight (g/mol) | 542.04 |
| MDL Number | MFCD00015318 |
| InChI Key | REQVOKPBMSCTNI-UHFFFAOYSA-L |
| PubChem CID | 131675641 |
| IUPAC Name | 3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride |
| SMILES | Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC291590010
|
Thermo Scientific Chemicals
291590010 |
1 g | Glass bottle |
Each for $73.34
|
|
||||
|
AC291590050
|
Thermo Scientific Chemicals
291590050 |
5 g | Glass bottle |
Each for $142.39
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15629-92-2 | |
| 542.04 | |
| REQVOKPBMSCTNI-UHFFFAOYSA-L | |
| 3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride |
| C27H26Cl2NiP2 | |
| MFCD00015318 | |
| 131675641 | |
| Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
| 15629-92-2 | |
| 100.0 | |
| 98.5% min. | |
| 99% | |
| C27H26Cl2NiP2 | |
| 1 g | |
| Solubility in water: insoluble | |
| Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 542.04 | |
| 542.04 | |
| Crystals or Powder |
| 98.5 | |
| 213.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00015318 | |
| 06,50; 15,122 | |
| REQVOKPBMSCTNI-UHFFFAOYSA-L | |
| 3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride | |
| 131675641 | |
| 99% | |
| [1,3-Bis(diphenylphosphino)propane]nickel(II) chloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON
GHS Signal Word: Danger
RUO – Research Use Only