missing translation for 'onlineSavingsMsg'
Learn More

[1,3-Bis(diphenylphosphino)propane]nickel(II) chloride, 98+%

Catalog No. AC291590010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC291590010 1 g Glass bottle
AC291590050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC291590010 Supplier Thermo Scientific Chemicals Supplier No. 291590010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15629-92-2 | C27H26Cl2NiP2 | 542.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15629-92-2
Molecular Formula C27H26Cl2NiP2
Molecular Weight (g/mol) 542.04
MDL Number MFCD00015318
InChI Key REQVOKPBMSCTNI-UHFFFAOYSA-L
PubChem CID 131675641
IUPAC Name 3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride
SMILES Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 213.0°C
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Fieser 06,50; 15,122
Solubility Information Solubility in water: insoluble
Formula Weight 542.04
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material [1,3-Bis(diphenylphosphino)propane]nickel(II) chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON

RUO – Research Use Only