missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(diphenylphosphino)propane, 97%

Catalog No. AC310050050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC310050050 5 g Glass bottle
AC310050250 25 g Glass bottle
AC310051000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC310050050 Supplier Thermo Scientific Chemicals Supplier No. 310050050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6737-42-4 | C27H26P2 | 412.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6737-42-4
Molecular Formula C27H26P2
Molecular Weight (g/mol) 412.45
MDL Number MFCD00003050
InChI Key LVEYOSJUKRVCCF-UHFFFAOYSA-N
Synonym 1,3-bis diphenylphosphino propane, dppp, trimethylenebis diphenylphosphine, phosphine, 1,3-propanediylbis diphenyl, 3-diphenylphosphanyl propyl diphenylphosphane, 3-diphenylphosphanylpropyl diphenyl phosphane, 1,3-bis diphenyphosphino propane, 1,3-bis diphenylphosphino propane dppp, bis 1,3-diphenylphosphino propane, 1,3-bis-diphenylphosphino propane
PubChem CID 81219
SMILES C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 57.0°C to 63.0°C
Elemental Analysis 96.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5)2P(CH2)3P(C6H5)2
Quantity 5 g
Merck Index 15,1254
Solubility Information Solubility in water: insoluble
IUPAC Name 3-diphenylphosphanylpropyl(diphenyl)phosphane
Formula Weight 412.46
Percent Purity 97%
Physical Form Powder or Granules
Chemical Name or Material 1, 3-Bis(diphenylphosphino)propane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 229-791-2

RUO – Research Use Only