missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[5-(4-tert-butylphenyl)-2-[1,3,4]oxadiazolyl]benzene 97.0+%, TCI America™

Catalog No. p-7123432
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4252200MG 200 mg
B42521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4252200MG Supplier TCI America Supplier No. B4252200MG

Chemical Identifiers

CAS 138372-67-5
Molecular Formula C30H30N4O2
Molecular Weight (g/mol) 478.60
MDL Number MFCD03093214
InChI Key FQJQNLKWTRGIEB-UHFFFAOYSA-N
Synonym 1,3-Bis[5-(4-tert-butylphenyl)-1,3,4-oxadiazol-2-yl]benzene, OXD-7
PubChem CID 15320492
IUPAC Name 2-(4-tert-butylphenyl)-5-{3-[5-(4-tert-butylphenyl)-1,3,4-oxadiazol-2-yl]phenyl}-1,3,4-oxadiazole
SMILES CC(C)(C)C1=CC=C(C=C1)C1=NN=C(O1)C1=CC(=CC=C1)C1=NN=C(O1)C1=CC=C(C=C1)C(C)(C)C

Specifications

Melting Point 241°C
Color White
Quantity 200 mg
Formula Weight 478.60
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[5-(4-tert-butylphenyl)-2-[1,3,4]oxadiazolyl]benzene