missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[4-(trifluoromethyl)phenyl]urea 98.0+%, TCI America™

Catalog No. p-7123591
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4518200MG 200 mg
B45181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4518200MG Supplier TCI America Supplier No. B4518200MG
Only null left

Chemical Identifiers

CAS 1960-88-9
Molecular Formula C15H10F6N2O
Molecular Weight (g/mol) 348.248
MDL Number MFCD01908917
InChI Key PEXQIAHZLFTVJU-UHFFFAOYSA-N
Synonym 1,3-bis 4-trifluoromethyl phenyl urea, maybridge4_001077, 1,3-bis 4-trifluoromethylphenyl urea, n,n'-bis-4-trifluoromethylphenyl-urea, urea, n,n'-bis 4-trifluoromethyl phenyl
PubChem CID 472928
IUPAC Name 1,3-bis[4-(trifluoromethyl)phenyl]urea
SMILES C1=CC(=CC=C1C(F)(F)F)NC(=O)NC2=CC=C(C=C2)C(F)(F)F

Specifications

Melting Point 227°C
Color White
Quantity 200 mg
Formula Weight 348.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[4-(trifluoromethyl)phenyl]urea