missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[4-(trifluoromethyl)phenyl]thiourea 97.0+%, TCI America™

$117.43 - $404.51

Chemical Identifiers

CAS 1744-07-6
Molecular Formula C15H10F6N2S
Molecular Weight (g/mol) 364.309
InChI Key ABYZUZQRUYXHCW-UHFFFAOYSA-N
PubChem CID 19743454
IUPAC Name 1,3-bis[4-(trifluoromethyl)phenyl]thiourea
SMILES C1=CC(=CC=C1C(F)(F)F)NC(=S)NC2=CC=C(C=C2)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B46111G
View Documents
TCI America
B46111G
1 g N/A
Only null left
 
B46115G
View Documents
TCI America
B46115G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1744-07-6
364.309
19743454
C1=CC(=CC=C1C(F)(F)F)NC(=S)NC2=CC=C(C=C2)C(F)(F)F
C15H10F6N2S
ABYZUZQRUYXHCW-UHFFFAOYSA-N
1,3-bis[4-(trifluoromethyl)phenyl]thiourea

Specifications

1744-07-6
Yellow
1 g
C1=CC(=CC=C1C(F)(F)F)NC(=S)NC2=CC=C(C=C2)C(F)(F)F
364.309
364.31
Crystalline Powder
158°C
C15H10F6N2S
ABYZUZQRUYXHCW-UHFFFAOYSA-N
1,3-bis[4-(trifluoromethyl)phenyl]thiourea
19743454
≥97.0% (HPLC,N)
1,3-Bis[4-(trifluoromethyl)phenyl]thiourea
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'