missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[4-(trifluoromethyl)phenyl]thiourea 97.0+%, TCI America™

Catalog No. B46111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46111G 1 g
B46115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46111G Supplier TCI America Supplier No. B46111G
Only null left

Chemical Identifiers

CAS 1744-07-6
Molecular Formula C15H10F6N2S
Molecular Weight (g/mol) 364.309
InChI Key ABYZUZQRUYXHCW-UHFFFAOYSA-N
PubChem CID 19743454
IUPAC Name 1,3-bis[4-(trifluoromethyl)phenyl]thiourea
SMILES C1=CC(=CC=C1C(F)(F)F)NC(=S)NC2=CC=C(C=C2)C(F)(F)F

Specifications

Melting Point 158°C
Color Yellow
Quantity 1 g
Formula Weight 364.31
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[4-(trifluoromethyl)phenyl]thiourea