missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(4-methoxyphenyl)urea 98.0+%, TCI America™
$290.79 - $1002.86
Chemical Identifiers
| CAS | 1227-44-7 |
|---|---|
| Molecular Formula | C15H16N2O3 |
| Molecular Weight (g/mol) | 272.304 |
| MDL Number | MFCD00025783 |
| InChI Key | CPSIMDUNVYMPAH-UHFFFAOYSA-N |
| PubChem CID | 136951 |
| IUPAC Name | 1,3-bis(4-methoxyphenyl)urea |
| SMILES | COC1=CC=C(C=C1)NC(=O)NC2=CC=C(C=C2)OC |
Chemical Identifiers
| 1227-44-7 | |
| 272.304 | |
| CPSIMDUNVYMPAH-UHFFFAOYSA-N | |
| 1,3-bis(4-methoxyphenyl)urea |
| C15H16N2O3 | |
| MFCD00025783 | |
| 136951 | |
| COC1=CC=C(C=C1)NC(=O)NC2=CC=C(C=C2)OC |
Specifications
| 1227-44-7 | |
| C15H16N2O3 | |
| 1 g | |
| COC1=CC=C(C=C1)NC(=O)NC2=CC=C(C=C2)OC | |
| 272.304 | |
| 272.30 | |
| Crystalline Powder |
| 241°C | |
| MFCD00025783 | |
| CPSIMDUNVYMPAH-UHFFFAOYSA-N | |
| 1,3-bis(4-methoxyphenyl)urea | |
| 136951 | |
| ≥98.0% (HPLC,N) | |
| 1,3-Bis(4-methoxyphenyl)urea |
Safety and Handling
TSCA : No