missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-methoxyphenyl)urea 98.0+%, TCI America™

Catalog No. B44831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44831G 1 g
B44835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44831G Supplier TCI America Supplier No. B44831G

Chemical Identifiers

CAS 1227-44-7
Molecular Formula C15H16N2O3
Molecular Weight (g/mol) 272.304
MDL Number MFCD00025783
InChI Key CPSIMDUNVYMPAH-UHFFFAOYSA-N
PubChem CID 136951
IUPAC Name 1,3-bis(4-methoxyphenyl)urea
SMILES COC1=CC=C(C=C1)NC(=O)NC2=CC=C(C=C2)OC

Specifications

Melting Point 241°C
Quantity 1 g
Formula Weight 272.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(4-methoxyphenyl)urea